missing translation for 'onlineSavingsMsg'
Learn More

Hesperetin, MilliporeSigma™ Supelco™

Catalog No. 111015250
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015250 1 g
111015251 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015250 Supplier Merck Emd Millipore Supplier No. H41251G
Only null left

Hesperetin is an aglycon form of flavonoid, which can be found in peels of citrus fruits.

Featured Industries: Food and Beverages

Specifications

CAS 69097-99-0
Physical Form Neat
Linear Formula C16H14O6
MDL Number MFCD00016956
Synonym 3′,5,7-Trihydroxy-4′-methoxyflavanone; 5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-4-chromanone
Type Analytical Standard
Quantity 1 g
Formula Weight 302.28
Grade Analytical Standard