missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Heraguard™ ECO Clean Bench Accessories

Integrate Thermo Scientific™ Heraguard™ ECO Clean Bench Accessories with the ECO clean bench to increase the flexibility and enhance operations.

Supplier:  Thermo Scientific™ 50045959

 View more versions of this product

Catalog No. 01670814


Only null left

Description

Description

Integrate Thermo Scientific™ Heraguard™ ECO Clean Bench Accessories with the ECO clean bench to increase the flexibility and enhance operations. Options include media valves, solenoid valve, stainless-steel work surface wraps, side window modifications, UV and UV night covers, windscreen protection kits and additional accessories.

† Due to DC motors, HERAguard ECO clean benches use less energy to operate so there is less heat emitted into the lab than traditional AC motor clean benches. Based on data from internal testing and The University of Michigan Field Study, published in American Biotechnology Laboratory. 

Specifications

Specifications

CD3/CD27
Heraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95
M2AB, DEN-3, DEN-3
PBS with 0.2% carrier protein and 0.08% sodium azide
P04234, P07766, P09693, P26842
CD27, CD3D, CD3E, CD3G
human thymocytes
100 Tests
RUO
Primary
Human
Purified
Field Installed
Flow Cytometry
Cocktail
FITC/PE
CD3E
CD3e, CD3 epsilon, FLJ18683, T3E, TCRE, CD3-epsilon, CD3e antigen, epsilon polypeptide TiT3 complex, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TNFRSF7, CD27 antigen, CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T-cell activation antigen CD27, tumor necrosis factor receptor superfamily member 7
Murine
Protein A/G, Protein A/G
Non-combustible gas G3/8 gas connection (green) with long neck for installation in side walls
Non-combustible Gas Connection, Antibody
915, 915, 916, 916, 917, 917, 939, 939
Store at 4°C, do not freeze
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions