Learn More
Thermo Scientific™ Heraguard™ ECO Clean Bench Accessories
Integrate Thermo Scientific™ Heraguard™ ECO Clean Bench Accessories with the ECO clean bench to increase the flexibility and enhance operations.
Supplier: Thermo Scientific™ 50044678
Description
Integrate Thermo Scientific™ Heraguard™ ECO Clean Bench Accessories with the ECO clean bench to increase the flexibility and enhance operations. Options include media valves, solenoid valve, stainless-steel work surface wraps, side window modifications, UV and UV night covers, windscreen protection kits and additional accessories.
† Due to DC motors, HERAguard ECO clean benches use less energy to operate so there is less heat emitted into the lab than traditional AC motor clean benches. Based on data from internal testing and The University of Michigan Field Study, published in American Biotechnology Laboratory.
Specifications
| CD3/CD26 | |
| Heraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95; Herasafe 2030i 0.9, 1.2, 1.5, 1.8; Maxisafe 2030i 0.9, 1.2, 1.5, 1.8 | |
| M2AB, EUG-6, EUG-6 | |
| PBS with 0.2% carrier protein and 0.08% sodium azide | |
| P04234, P07766, P09693, P27487 | |
| CD3D, CD3E, CD3G, DPP4 | |
| human thymocytes | |
| 50 Tests | |
| RUO | |
| Primary | |
| Human | |
| Purified | |
| Field Installed |
| Flow Cytometry | |
| Cocktail | |
| FITC/PE | |
| CD3E | |
| CD3e, CD3 epsilon, FLJ18683, T3E, TCRE, CD3-epsilon, CD3e antigen, epsilon polypeptide TiT3 complex, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, DPP4, ADABP, ADCP2, CD26, DPP-IV, TP103, ADCP-2, DPP IV, T-cell activation antigen CD26, adenosine deaminase complexing protein 2, dipeptidyl peptidase 4, dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidylpeptidase IV CD26, adenosine deaminase complexing protein 2 | |
| Murine | |
| Protein A/G, Protein A/G | |
| Vacuum tap G3/8 connection (grey) with long neck for installation in side walls | |
| Vacuum Tap Connection, Antibody | |
| 1803, 1803, 915, 915, 916, 916, 917, 917 | |
| Store at 4°C, do not freeze | |
| IgG1, IgG2a |