missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Heraguard™ ECO Clean Bench Accessories

Catalog No. 01670811
Change view
Click to view available options
Product Type:
Cover
General Accessories
Lead Through, 10 to 13 mm ID
Lead Through, 3 mm ID
Non-combustible Gas Connection, Antibody
UV Options
Vacuum Tap Connection, Antibody
Water Tap Connection, Antibody
Wind Screen Protection Kit
For Use With (Equipment):
Heraguard ECO 0.9
Heraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95
Heraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95; Herasafe 2030i 0.9, 1.2, 1.5, 1.8; Maxisafe 2030i 0.9, 1.2, 1.5, 1.8
Heraguard ECO 1.2
Heraguard ECO 1.2, 1.5,1.8, 1.8/95
Heraguard ECO 1.5
Heraguard ECO 1.8
Heraguard ECO 1.8/95
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Equipment)
01670811 Water Tap Connection, Antibody Heraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95
13261522 Wind Screen Protection Kit Heraguard ECO 0.9
13998029 General Accessories Heraguard ECO 0.9
13998030 General Accessories Heraguard ECO 1.2, 1.5,1.8, 1.8/95
13261527 Cover Heraguard ECO 1.2
13998025 Lead Through, 10 to 13 mm ID Heraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95
01670814 Non-combustible Gas Connection, Antibody Heraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95
13261523 Wind Screen Protection Kit Heraguard ECO 1.2
13261526 Cover Heraguard ECO 0.9
13261524 Wind Screen Protection Kit Heraguard ECO 1.5
01670813 Vacuum Tap Connection, Antibody Heraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95; Herasafe 2030i 0.9, 1.2, 1.5, 1.8; Maxisafe 2030i 0.9, 1.2, 1.5, 1.8
13261516 UV Options Heraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95
13261529 Cover Heraguard ECO 1.8
13998026 Lead Through, 3 mm ID Heraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95
13261528 Cover Heraguard ECO 1.5
13261525 Wind Screen Protection Kit Heraguard ECO 1.8
13261530 Cover Heraguard ECO 1.8/95
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. 01670811 Supplier Thermo Scientific™ Supplier No. 50044679

Integrate Thermo Scientific™ Heraguard™ ECO Clean Bench Accessories with the ECO clean bench to increase the flexibility and enhance operations.

Integrate Thermo Scientific™ Heraguard™ ECO Clean Bench Accessories with the ECO clean bench to increase the flexibility and enhance operations. Options include media valves, solenoid valve, stainless-steel work surface wraps, side window modifications, UV and UV night covers, windscreen protection kits and additional accessories.

† Due to DC motors, HERAguard ECO clean benches use less energy to operate so there is less heat emitted into the lab than traditional AC motor clean benches. Based on data from internal testing and The University of Michigan Field Study, published in American Biotechnology Laboratory. 

Specifications

Antigen CD3/CD25
Applications Flow Cytometry
For Use With (Equipment) Heraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95
Classification Cocktail
Clone M2AB, ITYV, ITYV
Conjugate FITC/PE
Formulation PBS with 0.2% carrier protein and 0.08% sodium azide
Gene CD3D
Gene Accession No. P01589, P04234, P07766, P09693
Gene Alias CD3e, CD3 epsilon, FLJ18683, T3E, TCRE, CD3-epsilon, CD3e antigen, epsilon polypeptide TiT3 complex, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, IL-2R alpha, IL2 RA antibody, Il2ra antibody, Interleukin 2 receptor alpha chain antibody, TAC antigen antibody, IL-2R alpha chain
Gene Symbols CD3D, CD3E, CD3G, IL2RA
Host Species Murine
Immunogen human thymocytes and PHA-activated human T-lymphocytes
Purification Method Protein A/G, Protein A/G
Quantity 50 Tests
Description Water tap G3/8 connection (Black) with long neck for installation in side walls
Regulatory Status RUO
Product Type Water Tap Connection, Antibody
Primary or Secondary Primary
Gene ID (Entrez) 3559, 3559, 915, 915, 916, 916, 917, 917
Target Species Human
Content And Storage Store at 4°C, do not freeze
Form Purified
Isotype IgG1
Configuration Options Field Installed
Show More Show Less