missing translation for 'onlineSavingsMsg'
Learn More

Heptyltriphenylphosphonium Bromide 98.0+%, TCI America™

Catalog No. H054525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H054525G 25 g
H0545500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H054525G Supplier TCI America Supplier No. H054525G

Chemical Identifiers

CAS 13423-48-8
Molecular Formula C25H30BrP
Molecular Weight (g/mol) 441.39
MDL Number MFCD00050249
InChI Key WCZSOHSGMBVYFW-UHFFFAOYSA-M
Synonym heptyltriphenylphosphonium bromide, n-heptyltriphenylphosphonium bromide, heptyltriphenylphosphanium bromide, heptyl triphenyl phosphanium bromide, heptyl triphenyl phosphonium bromide, n-heptyl-triphenylphosphonium bromide, 1-heptyl triphenylphosphonium bromide, phosphonium, heptyltriphenyl-, bromide, c7h15pph3br;, acmc-209bu9
PubChem CID 2724567
IUPAC Name heptyltriphenylphosphanium bromide
SMILES [Br-].CCCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 176°C
Color White
Quantity 25 g
Formula Weight 441.39
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Heptyltriphenylphosphonium Bromide