missing translation for 'onlineSavingsMsg'
Learn More

Heptylbenzene 97.0+%, TCI America™

Catalog No. p-7130309
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H004625ML 25 mL
H0046500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H004625ML Supplier TCI America Supplier No. H004625ML
Only null left

Chemical Identifiers

CAS 1078-71-3
Molecular Formula C13H20
Molecular Weight (g/mol) 176.303
MDL Number MFCD00009545
InChI Key LBNXAWYDQUGHGX-UHFFFAOYSA-N
Synonym 1-phenylheptane, benzene, heptyl, n-heptylbenzene, heptane, 1-phenyl, heptane, 1 or 7-phenyl, unii-12uc21c7sn, phenylheptane, heptyl-benzene, heptane, phenyl
PubChem CID 14115
IUPAC Name heptylbenzene
SMILES CCCCCCCC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 235°C
Quantity 25 mL
UN Number 3082
Formula Weight 176.30
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Heptylbenzene
EINECSNumber (3)-0021
TSCA Yes