missing translation for 'onlineSavingsMsg'
Learn More

Heptylamine 98.0+%, TCI America™

Catalog No. H004525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H004525ML 25 mL
H0045500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H004525ML Supplier TCI America Supplier No. H004525ML

Chemical Identifiers

CAS 111-68-2
Molecular Formula C7H17N
Molecular Weight (g/mol) 115.22
MDL Number MFCD00008244
InChI Key WJYIASZWHGOTOU-UHFFFAOYSA-N
Synonym heptylamine, 1-aminoheptane, 1-heptanamine, n-heptylamine, 1-heptylamine, heptanamine, heptyl-amine, heptyl amine, dsstox_cid_681
PubChem CID 8127
IUPAC Name heptan-1-amine
SMILES CCCCCCCN

Specifications

Color Yellow
Boiling Point 157°C
Quantity 25 mL
UN Number 2734
Formula Weight 115.22
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Heptylamine
EINECSNumber (2)-0133
RTECSNumber MK0600000
TSCA Yes