accessibility menu, dialog, popup

UserName

CAS RN 111-68-2

IUPAC Name: heptan-1-amine
Synonyms: heptylamine
Molecular Weight (g/mol): 115.22
Molecular Formula: C7H17N
InChi Key: WJYIASZWHGOTOU-UHFFFAOYSA-N
SMILES: CCCCCCCN
Boiling Point 
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)