missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptyl p-Toluenesulfonate 98.0+%, TCI America™
$226.48 - $2278.25
Chemical Identifiers
| CAS | 24767-82-6 |
|---|---|
| Molecular Formula | C14H22O3S |
| Molecular Weight (g/mol) | 270.387 |
| MDL Number | MFCD00059442 |
| InChI Key | BQPVBPLMUCJNOX-UHFFFAOYSA-N |
| Synonym | p-Toluenesulfonic Acid Heptyl Ester |
| PubChem CID | 599295 |
| IUPAC Name | heptyl 4-methylbenzenesulfonate |
| SMILES | CCCCCCCOS(=O)(=O)C1=CC=C(C=C1)C |
Chemical Identifiers
| 24767-82-6 | |
| 270.387 | |
| BQPVBPLMUCJNOX-UHFFFAOYSA-N | |
| 599295 | |
| CCCCCCCOS(=O)(=O)C1=CC=C(C=C1)C |
| C14H22O3S | |
| MFCD00059442 | |
| p-Toluenesulfonic Acid Heptyl Ester | |
| heptyl 4-methylbenzenesulfonate |
Specifications
| 24767-82-6 | |
| 201°C | |
| MFCD00059442 | |
| p-Toluenesulfonic Acid Heptyl Ester | |
| CCCCCCCOS(=O)(=O)C1=CC=C(C=C1)C | |
| 270.387 | |
| 270.39 | |
| Liquid |
| Yellow | |
| C14H22O3S | |
| 25 g | |
| BQPVBPLMUCJNOX-UHFFFAOYSA-N | |
| heptyl 4-methylbenzenesulfonate | |
| 599295 | |
| ≥98.0% (GC) | |
| Heptyl p-Toluenesulfonate |
Safety and Handling
TSCA : No