accessibility menu, dialog, popup

UserName

CAS RN 24767-82-6

H3COSOOCH3

CAS RN 24767-82-6

IUPAC Name: heptyl 4-methylbenzene-1-sulfonate
Synonyms: heptyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 270.39
Molecular Formula: C14H22O3S
InChi Key: BQPVBPLMUCJNOX-UHFFFAOYSA-N
SMILES: CCCCCCCOS(=O)(=O)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results