missing translation for 'onlineSavingsMsg'
Learn More

Heptyl p-Toluenesulfonate 98.0+%, TCI America™

Catalog No. T0884500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0884500G 500 g
T088425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0884500G Supplier TCI America Supplier No. T0884500G
Only null left

Chemical Identifiers

CAS 24767-82-6
Molecular Formula C14H22O3S
Molecular Weight (g/mol) 270.387
MDL Number MFCD00059442
InChI Key BQPVBPLMUCJNOX-UHFFFAOYSA-N
Synonym p-Toluenesulfonic Acid Heptyl Ester
PubChem CID 599295
IUPAC Name heptyl 4-methylbenzenesulfonate
SMILES CCCCCCCOS(=O)(=O)C1=CC=C(C=C1)C

Specifications

Color Yellow
Boiling Point 201°C
Quantity 500 g
Formula Weight 270.39
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Heptyl p-Toluenesulfonate