missing translation for 'onlineSavingsMsg'
Learn More

Heptyl Isocyanate 97.0+%, TCI America™

Catalog No. I050510ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I050510ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I050510ML Supplier TCI America Supplier No. I050510ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4747-81-3
Molecular Formula C8H15NO
Molecular Weight (g/mol) 141.21
MDL Number MFCD00144892
InChI Key RFXBSYPBSRSQDU-UHFFFAOYSA-N
Synonym heptyl isocyanate, n-heptyl isocyanate, 1-heptyl isocyanate, heptane, 1-isocyanato, n-heptylisocyanate, heptylisocyanate, acmc-209k9f, isocyanic acid heptyl ester, 1-isocyanatoheptan-2-yl
PubChem CID 138341
IUPAC Name 1-isocyanatoheptane
SMILES CCCCCCCN=C=O

Specifications

Color Yellow
Boiling Point 184°C
Quantity 10 mL
UN Number 2206
Formula Weight 141.21
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Heptyl Isocyanate