accessibility menu, dialog, popup

UserName

H3CNCO

CAS RN 4747-81-3

IUPAC Name: 1-isocyanatoheptane
Synonyms: 1-isocyanatoheptane
Molecular Weight (g/mol): 141.21
Molecular Formula: C8H15NO
InChi Key: RFXBSYPBSRSQDU-UHFFFAOYSA-N
SMILES: CCCCCCCN=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results