missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptyl Acetate 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | -50°C |
| Color | Colorless |
| Boiling Point | 192°C |
| Quantity | 500 mL |
| Formula Weight | 158.24 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Heptyl Acetate |
Safety and Handling
| EINECSNumber | (2)-0735 |
| RTECSNumber | AH9901000 |
| TSCA | Yes |