accessibility menu, dialog, popup

UserName

H3COCH3O

CAS RN 112-06-1

IUPAC Name: heptyl acetate
Synonyms: heptyl acetate
Molecular Weight (g/mol): 158.24
Molecular Formula: C9H18O2
InChi Key: ZCZSIDMEHXZRLG-UHFFFAOYSA-N
SMILES: CCCCCCCOC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results