missing translation for 'onlineSavingsMsg'
Learn More

Heptyl Acetate 99.0+%, TCI America™

Catalog No. A090425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A090425ML 25 mL
A0904500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A090425ML Supplier TCI America Supplier No. A090425ML

Chemical Identifiers

CAS 112-06-1
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.24
MDL Number MFCD00027311
InChI Key ZCZSIDMEHXZRLG-UHFFFAOYSA-N
Synonym Acetic Acid Heptyl Ester
PubChem CID 8159
IUPAC Name heptyl acetate
SMILES CCCCCCCOC(C)=O

Specifications

Melting Point -50°C
Color Colorless
Boiling Point 192°C
Quantity 25 mL
Formula Weight 158.24
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Heptyl Acetate
EINECSNumber (2)-0735
RTECSNumber AH9901000
TSCA Yes