missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptyl 4-Hydroxybenzoate 98.0+%, TCI America™
Supplier: TCI America H0212100G
Specifications
| Heptyl 4-Hydroxybenzoate | |
| 47°C | |
| C14H20O3 | |
| 100 g | |
| ZTJORNVITHUQJA-UHFFFAOYSA-N | |
| heptyl 4-hydroxybenzoate | |
| 14138 | |
| 236.31 | |
| Crystalline Powder |
| 1085-12-7 | |
| White | |
| MFCD00016481 | |
| heptylparaben, heptyl paraben, heptyl p-hydroxybenzoate, n-heptyl 4-hydroxybenzoate, nipaheptyl, staypro ws 7, n-heptyl p-hydroxybenzoate, benzoic acid, 4-hydroxy-, heptyl ester, p-hydroxybenzoic acid heptyl ester, p-oxybenzoesaureheptylester | |
| CCCCCCCOC(=O)C1=CC=C(O)C=C1 | |
| 236.31 | |
| CHEBI:34788 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 1085-12-7 | |
| 236.31 | |
| ZTJORNVITHUQJA-UHFFFAOYSA-N | |
| 14138 | |
| heptyl 4-hydroxybenzoate |
| C14H20O3 | |
| MFCD00016481 | |
| heptylparaben, heptyl paraben, heptyl p-hydroxybenzoate, n-heptyl 4-hydroxybenzoate, nipaheptyl, staypro ws 7, n-heptyl p-hydroxybenzoate, benzoic acid, 4-hydroxy-, heptyl ester, p-hydroxybenzoic acid heptyl ester, p-oxybenzoesaureheptylester | |
| CHEBI:34788 | |
| CCCCCCCOC(=O)C1=CC=C(O)C=C1 |
Safety and Handling
EINECSNumber : (3)-1585
RTECSNumber : DH2202000
TSCA : Yes