missing translation for 'onlineSavingsMsg'
Learn More

Heptyl 4-Hydroxybenzoate 98.0+%, TCI America™

Catalog No. H021225G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H021225G 25 g
H0212100G 100 g
H0212500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H021225G Supplier TCI America Supplier No. H021225G

Chemical Identifiers

CAS 1085-12-7
Molecular Formula C14H20O3
Molecular Weight (g/mol) 236.31
MDL Number MFCD00016481
InChI Key ZTJORNVITHUQJA-UHFFFAOYSA-N
Synonym heptylparaben, heptyl paraben, heptyl p-hydroxybenzoate, n-heptyl 4-hydroxybenzoate, nipaheptyl, staypro ws 7, n-heptyl p-hydroxybenzoate, benzoic acid, 4-hydroxy-, heptyl ester, p-hydroxybenzoic acid heptyl ester, p-oxybenzoesaureheptylester
PubChem CID 14138
ChEBI CHEBI:34788
IUPAC Name heptyl 4-hydroxybenzoate
SMILES CCCCCCCOC(=O)C1=CC=C(O)C=C1

Specifications

Melting Point 47°C
Color White
Quantity 25 g
Formula Weight 236.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Heptyl 4-Hydroxybenzoate
EINECSNumber (3)-1585
RTECSNumber DH2202000
TSCA Yes