missing translation for 'onlineSavingsMsg'
Learn More

Heptanoyl Chloride 98.0+%, TCI America™

Supplier:  TCI America H0040500G

 View more versions of this product

Catalog No. H0040500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Heptanoyl Chloride
Yellow
C7H13ClO
500 g
enanthyl chloride, n-heptanoyl chloride, enanthic chloride, oenanthic chloride, heptanoic acid chloride, heptanoylchloride, enanthylchloride, enanthoyl chloride, c6h13cocl, heptanoyl chloride
CCCCCCC(Cl)=O
148.63
148.63
Liquid
2528-61-2
175°C
MFCD00000763
2920
UCVODTZQZHMTPN-UHFFFAOYSA-N
heptanoyl chloride
17313
≥98.0% (GC,T)

Chemical Identifiers

2528-61-2
148.63
UCVODTZQZHMTPN-UHFFFAOYSA-N
17313
CCCCCCC(Cl)=O
C7H13ClO
MFCD00000763
enanthyl chloride, n-heptanoyl chloride, enanthic chloride, oenanthic chloride, heptanoic acid chloride, heptanoylchloride, enanthylchloride, enanthoyl chloride, c6h13cocl, heptanoyl chloride
heptanoyl chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0632

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions