missing translation for 'onlineSavingsMsg'
Learn More

Heptanoyl Chloride 98.0+%, TCI America™

Catalog No. p-7130305
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H004025G 25 g
H0040100G 100 g
H0040500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H004025G Supplier TCI America Supplier No. H004025G
Only null left

Chemical Identifiers

CAS 2528-61-2
Molecular Formula C7H13ClO
Molecular Weight (g/mol) 148.63
MDL Number MFCD00000763
InChI Key UCVODTZQZHMTPN-UHFFFAOYSA-N
Synonym enanthyl chloride, n-heptanoyl chloride, enanthic chloride, oenanthic chloride, heptanoic acid chloride, heptanoylchloride, enanthylchloride, enanthoyl chloride, c6h13cocl, heptanoyl chloride
PubChem CID 17313
IUPAC Name heptanoyl chloride
SMILES CCCCCCC(Cl)=O

Specifications

Color Yellow
Boiling Point 175°C
Quantity 25 g
UN Number 2920
Formula Weight 148.63
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Heptanoyl Chloride
EINECSNumber (2)-0632
TSCA Yes