missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptanoic Anhydride 97.0+%, TCI America™
$63.38 - $384.00
Chemical Identifiers
| CAS | 626-27-7 |
|---|---|
| Molecular Formula | C14H26O3 |
| Molecular Weight (g/mol) | 242.359 |
| MDL Number | MFCD00009536 |
| InChI Key | DAPZDAPTZFJZTO-UHFFFAOYSA-N |
| Synonym | heptanoic anhydride, enanthic anhydride, heptanoic acid, anhydride, heptanoyl anhydride, n-heptanoic acid anhydride, n-heptanoic anhydride, heptanoic acid, 1,1'-anhydride, oenanthic anhydride, bisenanthic anhydride, enanthic acid anhydride |
| PubChem CID | 69376 |
| IUPAC Name | heptanoyl heptanoate |
| SMILES | CCCCCCC(=O)OC(=O)CCCCCC |
Chemical Identifiers
| 626-27-7 | |
| 242.359 | |
| DAPZDAPTZFJZTO-UHFFFAOYSA-N | |
| 69376 | |
| CCCCCCC(=O)OC(=O)CCCCCC |
| C14H26O3 | |
| MFCD00009536 | |
| heptanoic anhydride, enanthic anhydride, heptanoic acid, anhydride, heptanoyl anhydride, n-heptanoic acid anhydride, n-heptanoic anhydride, heptanoic acid, 1,1'-anhydride, oenanthic anhydride, bisenanthic anhydride, enanthic acid anhydride | |
| heptanoyl heptanoate |
Specifications
| 626-27-7 | |
| Yellow | |
| C14H26O3 | |
| 25 mL | |
| heptanoic anhydride, enanthic anhydride, heptanoic acid, anhydride, heptanoyl anhydride, n-heptanoic acid anhydride, n-heptanoic anhydride, heptanoic acid, 1,1'-anhydride, oenanthic anhydride, bisenanthic anhydride, enanthic acid anhydride | |
| CCCCCCC(=O)OC(=O)CCCCCC | |
| 242.359 | |
| 242.36 | |
| Liquid |
| -12°C | |
| 260°C | |
| MFCD00009536 | |
| 3265 | |
| DAPZDAPTZFJZTO-UHFFFAOYSA-N | |
| heptanoyl heptanoate | |
| 69376 | |
| ≥97.0% (T) | |
| Heptanoic Anhydride |
Safety and Handling
TSCA : Yes