missing translation for 'onlineSavingsMsg'
Learn More

Heptanoic Acid 98.0+%, TCI America™

Catalog No. H0030500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0030500ML 500 mL
H003025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0030500ML Supplier TCI America Supplier No. H0030500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111-14-8
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00004426
InChI Key MNWFXJYAOYHMED-UHFFFAOYSA-N
Synonym enanthic acid, oenanthic acid, n-heptanoic acid, enanthylic acid, heptoic acid, n-heptoic acid, n-heptylic acid, heptylic acid, oenanthylic acid, 1-hexanecarboxylic acid
PubChem CID 8094
ChEBI CHEBI:45571
IUPAC Name heptanoic acid
SMILES CCCCCCC(=O)O

Specifications

Melting Point -8°C
Color Colorless
Boiling Point 223°C
Quantity 500 mL
UN Number 3265
Formula Weight 130.19
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Heptanoic Acid
EINECSNumber (2)-0608
RTECSNumber MJ1575000
TSCA Yes