accessibility menu, dialog, popup

UserName

CAS RN 111-14-8

IUPAC Name: heptanoic acid
Synonyms: heptanoic acid heptansaeure N-heptanoic acid heptanoic acid (natural) oenanthylic acid enanthylic acid hexacid C-7 oenanthsaeure
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: MNWFXJYAOYHMED-UHFFFAOYSA-N
SMILES: CCCCCCC(O)=O
Boiling Point 
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)