Learn More
Heptanoic acid, 98+%
CAS: 111-14-8 | C7H14O2 | 130.187 g/mol
$127.44 - $443.95
Chemical Identifiers
| CAS | 111-14-8 |
|---|---|
| Molecular Formula | C7H14O2 |
| Molecular Weight (g/mol) | 130.187 |
| MDL Number | MFCD00004426 |
| InChI Key | MNWFXJYAOYHMED-UHFFFAOYSA-N |
| Synonym | enanthic acid, oenanthic acid, n-heptanoic acid, enanthylic acid, heptoic acid, n-heptoic acid, n-heptylic acid, heptylic acid, oenanthylic acid, 1-hexanecarboxylic acid |
| PubChem CID | 8094 |
| ChEBI | CHEBI:45571 |
| IUPAC Name | heptanoic acid |
| SMILES | CCCCCCC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA17704AP
|
Thermo Scientific Chemicals
A17704AP |
500 mL |
Each for $127.44
|
|
|||||
|
AAA177040F
|
Thermo Scientific Chemicals
A177040F |
2500 mL |
Each for $443.95
|
|
|||||
Description
Heptanoic acid is used as an organic building block for the synthesis of a variety of chemical compounds. It is involved in the esterification of steroids and used in the preparation of active pharmaceutical ingredients such as trenbolone enanthate, testosterone enanthate, drostanolone enanthate and methenolone enanthate. It is also used as an internal standard during gas chromatography analysis of the butyl esters of volatile acids. Its ester derivatives are used in the fragrance and flavor industries as well as cosmetics and industrial lubricants in aviation, refrigeration and automobile. It is used to prepare sodium heptanoate, which is used as a corrosion inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsHeptanoic acid is used as an organic building block for the synthesis of a variety of chemical compounds. It is involved in the esterification of steroids and used in the preparation of active pharmaceutical ingredients such as trenbolone enanthate, testosterone enanthate, drostanolone enanthate and methenolone enanthate. It is also used as an internal standard during gas chromatography analysis of the butyl esters of volatile acids. Its ester derivatives are used in the fragrance and flavor industries as well as cosmetics and industrial lubricants in aviation, refrigeration and automobile. It is used to prepare sodium heptanoate, which is used as a corrosion inhibitor.
Solubility
Miscible with acetone, ethanol, diethyl ether, dimethylformamide, dimethyl sulfoxide and nitric acid. Slightly soluble in carbon tetrachloride and water.
Notes
Incompatible with strong oxidizing agents, bases and reducing agents.
Chemical Identifiers
| 111-14-8 | |
| 130.187 | |
| MNWFXJYAOYHMED-UHFFFAOYSA-N | |
| 8094 | |
| heptanoic acid |
| C7H14O2 | |
| MFCD00004426 | |
| enanthic acid, oenanthic acid, n-heptanoic acid, enanthylic acid, heptoic acid, n-heptoic acid, n-heptylic acid, heptylic acid, oenanthylic acid, 1-hexanecarboxylic acid | |
| CHEBI:45571 | |
| CCCCCCC(=O)O |
Specifications
| 111-14-8 | |
| 0.917 | |
| 114°C (237°F) | |
| 1.423 | |
| MFCD00004426 | |
| UN3265 | |
| 14,4660 | |
| Miscible with acetone,ethanol,diethyl ether,dimethylformamide,dimethyl sulfoxide and nitric acid. Slightly soluble in carbon tetrachloride and water. | |
| CCCCCCC(=O)O | |
| 130.187 | |
| CHEBI:45571 | |
| ≥98% |
| -8°C | |
| 222°C to 223°C | |
| C7H14O2 | |
| CH3(CH2)5CO2H | |
| 500 mL | |
| 1744723 | |
| enanthic acid, oenanthic acid, n-heptanoic acid, enanthylic acid, heptoic acid, n-heptoic acid, n-heptylic acid, heptylic acid, oenanthylic acid, 1-hexanecarboxylic acid | |
| MNWFXJYAOYHMED-UHFFFAOYSA-N | |
| heptanoic acid | |
| 8094 | |
| 130.19 | |
| Heptanoic acid |
Safety and Handling
GHS H Statement
H314
Causes severe skin burns and eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 203-838-7
RTECSNumber : MJ1575000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only