missing translation for 'onlineSavingsMsg'
Learn More

Heptanoic acid, 98%

Catalog No. p-140502
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164172500 250 mL Glass bottle
AC164170010 1 L Glass bottle
AC164170025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC164172500 Supplier Thermo Scientific Chemicals Supplier No. 164172500
Only null left

CAS: 111-14-8 | C7H14O2 | 130.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 111-14-8
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00004426
InChI Key MNWFXJYAOYHMED-UHFFFAOYSA-N
Synonym enanthic acid, oenanthic acid, n-heptanoic acid, enanthylic acid, heptoic acid, n-heptoic acid, n-heptylic acid, heptylic acid, oenanthylic acid, 1-hexanecarboxylic acid
PubChem CID 8094
ChEBI CHEBI:45571
IUPAC Name heptanoic acid
SMILES CCCCCCC(=O)O

Specifications

Melting Point -10.5°C
Color White
Density 0.9100g/mL
Boiling Point 223.0°C to 223.4°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4210 to 1.4240
Linear Formula CH3(CH2)5CO2H
Quantity 250 mL
Beilstein 02,338
Merck Index 15,4695
Specific Gravity 0.91
Solubility Information Solubility in water: 0.24g/100mL (15°C). Other solubilities: soluble in alcohol, ether, dmf, chloroform, petrol, soluble in most organic solvents
Viscosity 3.40 mPa.s (30°C)
Formula Weight 130.19
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Heptanoic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 203-838-7
RTECSNumber MJ1575000
TSCA TSCA

RUO – Research Use Only