missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptane (Spectranalyzed™), Fisher Chemical
Description
EcoSafPak™ Système d’emballage écologique fabriqué à partir de matériaux 100 % recyclables par un fournisseur certifié SFI.
Spécifications
Spécifications
| Point de fusion | -91 °C |
| Absorbance | Test Réussi |
| Couleur | Incolore |
| Densité | 0.683g/cm³ |
| Quantité | 4 L |
| Point d’éclair | −4 °C (25 °F) |
| Désignation | Test Réussi |
| Spectre infrarouge | Test Réussi |
| Emballage | Bouteille en verre |
| Forme physique | Liquide |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Flammable liquid and vapor. Inhalation may cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air. NFPA Health:1 Flammability:3 Instability:0 |
| DOTInformation | Class 3: Flammable Liquid |