missing translation for 'onlineSavingsMsg'
Learn More

Heptane, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.0%, Honeywell Burdick & Jackson

Catalog No. p-7112063
Change view
Click to view available options
Quantity:
19 L
204 L
Packaging:
Metal Can
Metal Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001034 19 L Metal Can
6001035 204 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001034 Supplier Honeywell Chemicals Supplier No. PP20619
Only null left

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC

Specifications

CAS Max % 100.0000%
Density 0.684 g/cm3
Boiling Point 98.4°C
Quantity 19 L
Flash Point −4°C
Packaging Metal Can
UN Number UN1206
Beilstein 1730763
Water 0.01% max.
Solubility Information 0.03% in Water at 20°C
Formula Weight 100.21
Percent Purity >99%
Grade ACS Reagent
Chemical Name or Material Heptane, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.0%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Heptanes