missing translation for 'onlineSavingsMsg'
Learn More

Heptane, HPLC Grade, Spectrum™ Chemical

Catalog No. p-7157583
Change view
Click to view available options
Quantity:
4 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603108 4 L Amber Glass Bottle
18603110 4 x 4 L Amber Glass Bottle
18603111 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18603108 Supplier Spectrum Chemical Mfg Cor Supplier No. HP6624LT
Only null left

HP662, 142-82-5, C7H16

Spectrum™ Chemical Heptane, HPLC Grade is a straight-chain alkane applied in laboratories as a non-polar mobile phase carrier solvent in high pressure liquid chromatography. All HPLC solvents offered by Spectrum are glass distilled, submicron filtered, and undergo rigorous specification testing to insure accurate, repeatable results.

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
IUPAC Name heptane
SMILES CCCCCCC

Specifications

CAS Max % 100%
Quantity 4 L
Packaging Amber Glass Bottle
Residue after Evaporation 2 ppm
UN Number UN1206
Water 0.02%
Boiling Point 98.4°C
Percent Purity >99%
Grade High Purity
Chemical Name or Material Heptane