missing translation for 'onlineSavingsMsg'
Learn More

Heptane, For pesticide residue analysis, Solstice

Catalog No. 6000632
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000632 4 x 4 L
6001026 1 L
6001027 2.5 L
60046352 4 x 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6000632 Supplier Solstice Supplier No. 344954X4L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CHROMASOLV™, for pesticide residue analysis

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC

Specifications

Melting Point -91°C (literature)
CAS Max % 100.0000%
Density 0.684 g/mL at 25 °C(lit.)
Quantity 4 x 4 L
Flash Point −4°C
Packaging Glass Bottle
Linear Formula CH3(CH2)5CH3
UN Number UN1206
Boiling Point 98.2°C to 98.4°C
Formula Weight 100.2
Percent Purity >99%
Grade Pesticide
Chemical Name or Material Heptane, For pesticide residue analysis
Show More Show Less
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Heptanes
EINECSNumber 205-563-8
Recommended Storage Room Temp