missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptane, for HPLC, GC, pesticide residue analysis and spectrophotometry, >99.5%, Solstice
Specifications
Specifications
| Absorbance | 0.1 AU max. at 225 nm, 0.01 AU max. at 250 nm |
| CAS Max % | 100.0000% |
| Density | 0.684 g/cm3 |
| Quantity | 20 L |
| Flash Point | −4°C |
| Packaging | HDPE Nowpak™ |
| UN Number | UN1206 |
| Water | 0.01% max. |
| Beilstein | 1730763 |
| Boiling Point | 98.4°C |
| Show More |
Safety and Handling
| ShelfLife | 547 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Heptanes |