missing translation for 'onlineSavingsMsg'
Learn More

Heptane, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.0%, Solstice

Catalog No. 6001037
Change view
Click to view available options
Quantity:
19 L
4 x 4 L
204 L
Packaging:
Glass Bottle
Metal Can
Metal Drum
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001037 204 L Metal Drum
6001038 4 x 4 L Glass Bottle
6001039 4 x 4 L Poly Bottle
6001036 19 L Metal Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6001037 Supplier Solstice Supplier No. LP206204

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC

Specifications

CAS Max % 100.0000%
Density 0.684 g/cm3
Quantity 204 L
Flash Point −4°C
Packaging Metal Drum
UN Number UN1206
Water 0.01% max.
Beilstein 1730763
Boiling Point 98.4°C
Solubility Information 0.03% in Water at 20°C
Formula Weight 100.21
Percent Purity >99%
Grade ACS
Assay by GC Analysis n-Heptane
Chemical Name or Material Heptane, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.0%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Heptanes