missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptane, 99%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor H10054LTGL
Specifications
| Heptane | |
| 1 | |
| 4 L | |
| Amber Glass Bottle | |
| 100.2 | |
| Reagent |
| 142-82-5 | |
| 98.4°C | |
| To Pass Test | |
| 1e-05 | |
| 0.99 |
Chemical Identifiers
| 142-82-5 | |
| 100.2 |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.