missing translation for 'onlineSavingsMsg'
Learn More

Heptane, 99%, Spectrum™ Chemical

Catalog No. 18610793
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610793 200 L Drum
18610795 500 mL Amber Glass Bottle
18610791 1 L Amber Glass Bottle
18610794 4 L Amber Glass Bottle
18610792 20 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18610793 Supplier Spectrum Chemical Mfg Cor Supplier No. H1005200LTBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 142-82-5
Molecular Weight (g/mol) 100.2

Specifications

CAS Max % 1
Quantity 200 L
Identification To Pass Test
Packaging Drum
Residue after Evaporation 1e-05
Boiling Point 98.4°C
Percent Purity 0.99
Grade Reagent
Chemical Name or Material Heptane

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.