missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptane, ≥99.8% (GC), MilliporeSigma™ Supelco™
Description
Petroleum
Specifications
Specifications
| Ignition Point | 433°F |
| CAS | 142-82-5 |
| Vapor Pressure | 83 mmHg (37.7°C) |
| Vapor Density | 3.5 (vs. air) |
| Density | 0.684 g/mL (at 25°C) |
| Flash Point | 22°C |
| Physical Form | Neat |
| Linear Formula | CH3(CH2)5 CH3 |
| MDL Number | MFCD00009544 |
| Melting Point | -91°C |
| Show More |