missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptamethyldisilazane 96.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 147°C |
| Quantity | 25 mL |
| UN Number | 2924 |
| Formula Weight | 175.42 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Heptamethyldisilazane |
Safety and Handling
| TSCA | Yes |