accessibility menu, dialog, popup

UserName

CH3NSiH3CCH3H3CSiCH3CH3H3C

CAS RN 920-68-3

IUPAC Name: methylbis(trimethylsilyl)amine
Synonyms: methylbis(trimethylsilyl)amine
Molecular Weight (g/mol): 175.42
Molecular Formula: C7H21NSi2
InChi Key: ZSMNRKGGHXLZEC-UHFFFAOYSA-N
SMILES: CN([Si](C)(C)C)[Si](C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results