missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptakis-6-bromo-6-deoxy-beta-cyclodextrin, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 299005000
| Quantity | 500 mg |
|---|
Description
Chelation/Complexation reagent
Chemical Identifiers
| 53784-83-1 | |
| 1575.27 | |
| XNNRROJOWZLYCK-UHFFFAOYNA-N | |
| 11137055 | |
| OC1C(O)C2OC3OC(CBr)C(OC4OC(CBr)C(OC5OC(CBr)C(OC6OC(CBr)C(OC7OC(CBr)C(OC8OC(CBr)C(OC1OC2CBr)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O |
| C42H63Br7O28 | |
| MFCD00803289 | |
| heptakis 6-bromo-6-deoxy-beta-cyclodextrin | |
| 5,10,15,20,25,30,35-heptakis(bromomethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol |
Specifications
| Heptakis-6-bromo-6-deoxy-β-cyclodextrin | |
| 217°C to 221°C | |
| Glass bottle | |
| 500 mg | |
| heptakis 6-bromo-6-deoxy-beta-cyclodextrin | |
| XNNRROJOWZLYCK-UHFFFAOYNA-N | |
| 5,10,15,20,25,30,35-heptakis(bromomethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol | |
| 11137055 | |
| Powder |
| 53784-83-1 | |
| White | |
| C42H63Br7O28 | |
| MFCD00803289 | |
| Solubility in water: .. Other solubilities: soluble in dmf,dmso,pyridine | |
| OC1C(O)C2OC3OC(CBr)C(OC4OC(CBr)C(OC5OC(CBr)C(OC6OC(CBr)C(OC7OC(CBr)C(OC8OC(CBr)C(OC1OC2CBr)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O | |
| 1575.27 | |
| 1575.28 |