CAS RN 53784-83-1
CAS RN 53784-83-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Heptakis-6-bromo-6-deoxy-beta-cyclodextrin, Thermo Scientific™
CAS: 53784-83-1 Molecular Formula: C42H63Br7O28 Molecular Weight (g/mol): 1575.27 MDL Number: MFCD00803289 InChI Key: XNNRROJOWZLYCK-UHFFFAOYNA-N Synonym: heptakis 6-bromo-6-deoxy-beta-cyclodextrin PubChem CID: 11137055 IUPAC Name: 5,10,15,20,25,30,35-heptakis(bromomethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol SMILES: OC1C(O)C2OC3OC(CBr)C(OC4OC(CBr)C(OC5OC(CBr)C(OC6OC(CBr)C(OC7OC(CBr)C(OC8OC(CBr)C(OC1OC2CBr)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O