missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptafluorobutyric anhydride, ≥99.0%, MilliporeSigma™ Supelco™
Heptafluorobutyric anhydride (HFB) is a derivatizing reagent. It provides facile and sensitive way for detecting and elucidating structure of numerous alkyl-chain homologs.
$305.89 - $305.89
Chemical Identifiers
| MDL Number | MFCD00000432 |
|---|---|
| Synonym | HFBA; HFAA; HFBA; Perfluorobutyric anhydride |
Chemical Identifiers
| MFCD00000432 | |
| HFBA; HFAA; HFBA; Perfluorobutyric anhydride |
Specifications
| -43°C | |
| 1.674 g/mL (at 20°C) | |
| Ampule | |
| MFCD00000432 | |
| HFBA; HFAA; HFBA; Perfluorobutyric anhydride | |
| ≥99.0% |
| 336-59-4 | |
| 108°C to 110°C | |
| (CF3 CF2 CF2 CO)2 O | |
| 5 mL | |
| 410.06 | |
| For GC derivatization |