missing translation for 'onlineSavingsMsg'
Learn More

Heptafluorobutyric Acid 98.0+%, TCI America™

Catalog No. p-7130292
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H002425G 25 g
H0024100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H002425G Supplier TCI America Supplier No. H002425G
Only null left

Chemical Identifiers

CAS 375-22-4
Molecular Formula C4HF7O2
Molecular Weight (g/mol) 214.039
MDL Number MFCD00004171
InChI Key YPJUNDFVDDCYIH-UHFFFAOYSA-N
Synonym heptafluorobutyric acid, perfluorobutyric acid, heptafluorobutanoic acid, perfluorobutanoic acid, butanoic acid, heptafluoro, heptafluoro-1-butanoic acid, perfluoropropanecarboxylic acid, butyric acid, heptafluoro, kyselina heptafluormaselna, kyselina heptafluormaselna czech
PubChem CID 9777
ChEBI CHEBI:39426
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutanoic acid
SMILES C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O

Specifications

Melting Point -20°C
Color Yellow
Boiling Point 120°C
Quantity 25 g
UN Number 3265
Formula Weight 214.04
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Heptafluorobutyric Acid
EINECSNumber (2)-1182
RTECSNumber ET4025000
TSCA Yes