missing translation for 'onlineSavingsMsg'
Learn More

Heptadecanoic Acid 98.0+%, TCI America™

Supplier:  TCI America H0019100G

Quantity 100 g

 View more versions of this product

Catalog No. H0019100G


Only null left

Description

Description

Specifications

Chemical Identifiers

506-12-7
270.457
KEMQGTRYUADPNZ-UHFFFAOYSA-N
10465
heptadecanoic acid
C17H34O2
MFCD00002751
margaric acid, n-heptadecanoic acid, margarinic acid, n-heptadecylic acid, n-heptadecoic acid, heptadecylic acid, fatty acids, c16-18, unii-v987y9oz8l, normal-heptadecanoic acid, margarinate
CHEBI:32365
CCCCCCCCCCCCCCCCC(=O)O

Specifications

Heptadecanoic Acid
61°C
220°C
100 g
margaric acid, n-heptadecanoic acid, margarinic acid, n-heptadecylic acid, n-heptadecoic acid, heptadecylic acid, fatty acids, c16-18, unii-v987y9oz8l, normal-heptadecanoic acid, margarinate
CCCCCCCCCCCCCCCCC(=O)O
270.457
CHEBI:32365
≥98.0% (GC,T)
506-12-7
White
C17H34O2
MFCD00002751
KEMQGTRYUADPNZ-UHFFFAOYSA-N
heptadecanoic acid
10465
270.46
Crystalline Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0608

RTECSNumber : MI3850000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions