missing translation for 'onlineSavingsMsg'
Learn More

Heptadecanoic Acid 98.0+%, TCI America™

Catalog No. p-7130288
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H001925G 25 g
H0019100G 100 g
H0019500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H001925G Supplier TCI America Supplier No. H001925G

Chemical Identifiers

CAS 506-12-7
Molecular Formula C17H34O2
Molecular Weight (g/mol) 270.457
MDL Number MFCD00002751
InChI Key KEMQGTRYUADPNZ-UHFFFAOYSA-N
Synonym margaric acid, n-heptadecanoic acid, margarinic acid, n-heptadecylic acid, n-heptadecoic acid, heptadecylic acid, fatty acids, c16-18, unii-v987y9oz8l, normal-heptadecanoic acid, margarinate
PubChem CID 10465
ChEBI CHEBI:32365
IUPAC Name heptadecanoic acid
SMILES CCCCCCCCCCCCCCCCC(=O)O

Specifications

Melting Point 61°C
Color White
Boiling Point 220°C
Quantity 25 g
Formula Weight 270.46
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Heptadecanoic Acid
EINECSNumber (2)-0608
RTECSNumber MI3850000
TSCA Yes