missing translation for 'onlineSavingsMsg'
Learn More

Heptadecane 99.0+%, TCI America™

Catalog No. p-7130291
Change view
Click to view available options
Quantity:
25 g
250 g
Catalog No. Quantity
H002325G 25 g
H0023250G 250 g
2 options

Catalog No. H002325G

Supplier: TCI America H002325G

Only null left

Chemical Identifiers

CAS 629-78-7
Molecular Formula C17H36
Molecular Weight (g/mol) 240.475
MDL Number MFCD00009002
InChI Key NDJKXXJCMXVBJW-UHFFFAOYSA-N
Synonym n-heptadecane, heptadekan, unii-h7c0j39xum, hexadecane, methyl, h7c0j39xum, heptadecane, analytical standard, normal-heptadecane, heptadecane, dsstox_cid_27061, dsstox_rid_82078
PubChem CID 12398
ChEBI CHEBI:16148
IUPAC Name heptadecane
SMILES CCCCCCCCCCCCCCCCC

Specifications

Color White
Boiling Point 303°C
Quantity 25 g
Formula Weight 240.48
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Heptadecane
EINECSNumber (2)-0010
RTECSNumber MI3550000
TSCA Yes