missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptadecafluorononanoic Acid 95.0+%, TCI America™
$123.03 - $438.05
Chemical Identifiers
| CAS | 375-95-1 |
|---|---|
| Molecular Formula | C9HF17O2 |
| Molecular Weight (g/mol) | 464.078 |
| MDL Number | MFCD00039605 |
| InChI Key | UZUFPBIDKMEQEQ-UHFFFAOYSA-N |
| Synonym | perfluorononanoic acid, heptadecafluorononanoic acid, perfluoro-n-nonanoic acid, nonanoic acid, heptadecafluoro, perfluorononan-1-oic acid, pfna, heptadecafluoropelargonic acid, perfluornonansaeure, heptadecafluornonansaeure, heptadekafluornonansaeure |
| PubChem CID | 67821 |
| ChEBI | CHEBI:38397 |
| IUPAC Name | 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluorononanoic acid |
| SMILES | C(=O)(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O |
Chemical Identifiers
| 375-95-1 | |
| 464.078 | |
| UZUFPBIDKMEQEQ-UHFFFAOYSA-N | |
| 67821 | |
| 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluorononanoic acid |
| C9HF17O2 | |
| MFCD00039605 | |
| perfluorononanoic acid, heptadecafluorononanoic acid, perfluoro-n-nonanoic acid, nonanoic acid, heptadecafluoro, perfluorononan-1-oic acid, pfna, heptadecafluoropelargonic acid, perfluornonansaeure, heptadecafluornonansaeure, heptadekafluornonansaeure | |
| CHEBI:38397 | |
| C(=O)(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O |
Specifications
| 375-95-1 | |
| White-Yellow | |
| MFCD00039605 | |
| 1759 | |
| UZUFPBIDKMEQEQ-UHFFFAOYSA-N | |
| 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluorononanoic acid | |
| 67821 | |
| 464.08 | |
| Crystalline Powder |
| 65°C | |
| C9HF17O2 | |
| 5 g | |
| perfluorononanoic acid, heptadecafluorononanoic acid, perfluoro-n-nonanoic acid, nonanoic acid, heptadecafluoro, perfluorononan-1-oic acid, pfna, heptadecafluoropelargonic acid, perfluornonansaeure, heptadecafluornonansaeure, heptadekafluornonansaeure | |
| C(=O)(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O | |
| 464.078 | |
| CHEBI:38397 | |
| ≥95.0% (GC,T) | |
| Heptadecafluorononanoic Acid |
Safety and Handling
TSCA : Yes