CAS RN 375-95-1
CAS RN 375-95-1
IUPAC Name:
heptadecafluorononanoic acid
Synonyms:
perfluorononanoic acid heptadecafluornonansaeure perfluornonansaeure PFNA heptadekafluornonansaeure
Molecular Weight (g/mol):
464.08
Molecular Formula:
C9HF17O2
InChi Key:
UZUFPBIDKMEQEQ-UHFFFAOYSA-N
SMILES:
OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
1
–
1
of
1
results
1
Heptadecafluorononanoic Acid 95.0+%, TCI America™
CAS: 375-95-1 Molecular Formula: C9HF17O2 Molecular Weight (g/mol): 464.078 MDL Number: MFCD00039605 InChI Key: UZUFPBIDKMEQEQ-UHFFFAOYSA-N Synonym: perfluorononanoic acid,heptadecafluorononanoic acid,perfluoro-n-nonanoic acid,nonanoic acid, heptadecafluoro,perfluorononan-1-oic acid,pfna,heptadecafluoropelargonic acid,perfluornonansaeure,heptadecafluornonansaeure,heptadekafluornonansaeure PubChem CID: 67821 ChEBI: CHEBI:38397 IUPAC Name: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluorononanoic acid SMILES: C(=O)(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O