missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptadecafluoro-n-octyl Bromide 98.0+%, TCI America™
Supplier: TCI America H094625G
Specifications
| Heptadecafluoro-n-octyl Bromide | |
| Colorless | |
| C8BrF17 | |
| 25 g | |
| WTWWXOGTJWMJHI-UHFFFAOYSA-N | |
| 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane | |
| 9873 | |
| 498.96 | |
| Liquid |
| 423-55-2 | |
| 141°C | |
| MFCD00042082 | |
| perflubron, 1-bromoperfluorooctane, perfluorooctyl bromide, 1-bromoheptadecafluorooctane, oxygent, perfluoro-n-octyl bromide, imagent gi, perfluorooctylbromide, liquivent, perfluoroctyl bromide | |
| FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Br | |
| 498.97 | |
| CHEBI:38803 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 423-55-2 | |
| 498.97 | |
| WTWWXOGTJWMJHI-UHFFFAOYSA-N | |
| 9873 | |
| 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane |
| C8BrF17 | |
| MFCD00042082 | |
| perflubron, 1-bromoperfluorooctane, perfluorooctyl bromide, 1-bromoheptadecafluorooctane, oxygent, perfluoro-n-octyl bromide, imagent gi, perfluorooctylbromide, liquivent, perfluoroctyl bromide | |
| CHEBI:38803 | |
| FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Br |
Safety and Handling
RTECSNumber : RG8700000
TSCA : No