missing translation for 'onlineSavingsMsg'
Learn More

Heptadecafluoro-n-octyl Bromide 98.0+%, TCI America™

Catalog No. p-7130820
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09465G 5 g
H094625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09465G Supplier TCI America Supplier No. H09465G
Only null left

Chemical Identifiers

CAS 423-55-2
Molecular Formula C8BrF17
Molecular Weight (g/mol) 498.97
MDL Number MFCD00042082
InChI Key WTWWXOGTJWMJHI-UHFFFAOYSA-N
Synonym perflubron, 1-bromoperfluorooctane, perfluorooctyl bromide, 1-bromoheptadecafluorooctane, oxygent, perfluoro-n-octyl bromide, imagent gi, perfluorooctylbromide, liquivent, perfluoroctyl bromide
PubChem CID 9873
ChEBI CHEBI:38803
IUPAC Name 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Br

Specifications

Color Colorless
Boiling Point 141°C
Quantity 5 g
Formula Weight 498.96
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Heptadecafluoro-n-octyl Bromide
RTECSNumber RG8700000
TSCA No