missing translation for 'onlineSavingsMsg'
Learn More

Heptacosanoic Acid 98.0+%, TCI America™

Catalog No. H0971100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0971100MG 100 mg
H09711G 1 g
H09715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0971100MG Supplier TCI America Supplier No. H0971100MG
Only null left

Chemical Identifiers

CAS 7138-40-1
Molecular Formula C27H54O2
Molecular Weight (g/mol) 410.73
MDL Number MFCD00014029
InChI Key VXZBFBRLRNDJCS-UHFFFAOYSA-N
PubChem CID 23524
ChEBI CHEBI:78710
IUPAC Name heptacosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCC(O)=O

Specifications

Melting Point 86°C
Color White-Yellow
Quantity 100 mg
Formula Weight 410.73
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Heptacosanoic Acid
EINECSNumber (2)-0608
TSCA No