missing translation for 'onlineSavingsMsg'
Learn More

Heptachlor epoxide (Isomer B), Restek

Catalog No. 06719858
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06719858 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06719858 Supplier Chromatographic Supplier No. RK 32230

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1024-57-3
Molecular Formula C10H5Cl7O
Molecular Weight (g/mol) 389.299
InChI Key ZXFXBSWRVIQKOD-GOWUZIGHSA-N
PubChem CID 94865622
SMILES C12C(C(C3C1O3)Cl)C4(C(=C(C2(C4(Cl)Cl)Cl)Cl)Cl)Cl

Specifications

Quantity 1 mL
Chemical Name or Material Heptachlor epoxide (Isomer B)