missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals Inc HEPES Hemisodium Salt, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0215245180

 View more versions of this product

Catalog No. ICN15245180



Description

Description

Specifications

Specifications

HEPES Hemisodium Salt
White
C8H17N2NaO4S
100 g
RDZTWEVXRGYCFV-UHFFFAOYSA-M
sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
2724248
249.3
103404-87-1
1.005
MFCD00036463
hepes sodium salt, hepes hemisodium salt, sodium 2-4-2-hydroxyethyl piperazin-1-yl ethanesulfonate, unii-z9fto91o8a, 1-piperazineethanesulfonic acid, 4-2-hydroxyethyl-, monosodium salt, z9fto91o8a, sodium 4-2-hydroxyethyl piperazin-1-ylethanesulphonate, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid sodium salt, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid hemisodium salt
[Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1
260.28
CHEBI:46758
Powder

Chemical Identifiers

103404-87-1
260.28
RDZTWEVXRGYCFV-UHFFFAOYSA-M
2724248
sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
C8H17N2NaO4S
MFCD00036463
hepes sodium salt, hepes hemisodium salt, sodium 2-4-2-hydroxyethyl piperazin-1-yl ethanesulfonate, unii-z9fto91o8a, 1-piperazineethanesulfonic acid, 4-2-hydroxyethyl-, monosodium salt, z9fto91o8a, sodium 4-2-hydroxyethyl piperazin-1-ylethanesulphonate, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid sodium salt, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid hemisodium salt
CHEBI:46758
[Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1
Safety and Handling

Safety and Handling

Recommended Storage : Store at Room Temperature (15 to 30°C). Store Desiccated.

SDS
Documents

Documents

Promotions

Promotions